Potassium 9,9-dimethyl-9H-fluorene-2-trifluoroborate

Lieferant: Apollo Scientific

PC200175-1G PC200175-500MG PC200175-5G
APOSPC200175-1GEA 118 EUR
APOSPC200175-1G APOSPC200175-500MG APOSPC200175-5G
Potassium 9,9-dimethyl-9H-fluorene-2-trifluoroborate
Potassium 9,9-dimethyl-9H-fluorene-2-trifluoroborate
Formel: C₁₅H₁₃BF₃·K
Molecular Weight: 300,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20527106
CAS-Nummer: 2246944-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR