Potassium 4-hydroxyphenyltrifluoroborate
Lieferant: Apollo Scientific
PC200184-1G
PC200184-500MG
PC200184-5G
APOSPC200184-1GEA
139
EUR
APOSPC200184-1G
APOSPC200184-500MG
APOSPC200184-5G
Potassium 4-hydroxyphenyltrifluoroborate
Potassium 4-hydroxyphenyltrifluoroborate
Formel:
C₆H₅BF₃O·K Lagertemperatur: Raumtemperatur |
MDL:
MFCD09992974 CAS-Nummer: 1015082-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...