Potassium 5-bromo-2-thiophenetrifluoroborate
Lieferant: Apollo Scientific
Synonyme:
Potassium (5-bromothiophen-2-yl)trifluoroborate
PC200194-1G
PC200194-500MG
PC200194-5G
APOSPC200194-1GEA
147
EUR
APOSPC200194-1G
APOSPC200194-500MG
APOSPC200194-5G
Potassium 5-bromo-2-thiophenetrifluoroborate
Potassium 5-bromo-2-thiophenetrifluoroborate
Formel:
C₄H₂BBrF₃S·K Molecular Weight: 268,932 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17015269 CAS-Nummer: 1239370-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...