Methyl 5-amino-4-fluoro-1-methyl-1H-1,3-benzodiazole-6-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-amino-4-fluoro-1-methyl-1H-benzo[d]imidazole-6-carboxylate

PC200393-1G PC200393-500MG PC200393-250MG
APOSPC200393-1GEA 24.8 EUR
APOSPC200393-1G APOSPC200393-500MG APOSPC200393-250MG
Methyl 5-amino-4-fluoro-1-methyl-1H-1,3-benzodiazole-6-carboxylate
Methyl 5-amino-4-fluoro-1-methyl-1H-1,3-benzodiazole-6-carboxylate
Formel: C₁₀H₁₀FN₃O₂
Molecular Weight: 223,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26127252
CAS-Nummer: 918321-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR