2-Chloro-4-(4-fluorophenyl)-1,3,5-triazine
Lieferant: Apollo Scientific
|
Achtung
|
PC200401-1G
PC200401-5G
APOSPC200401-1GEA
183
EUR
APOSPC200401-1G
APOSPC200401-5G
2-Chloro-4-(4-fluorophenyl)-1,3,5-triazine
2-Chloro-4-(4-fluorophenyl)-1,3,5-triazine
Formel:
C₉H₅ClFN₃ Lagertemperatur: Kühlschrank |
MDL:
MFCD11052420 CAS-Nummer: 1053658-58-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...