1-Methyl-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine

Lieferant: Apollo Scientific

PC200413-250MG
APOSPC200413-250MGEA 217 EUR
APOSPC200413-250MG
1-Methyl-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine
1-Methyl-4-[3-(trifluoromethyl)pyridin-2-yl]piperazine
Formel: C₁₁H₁₄F₃N₃
Molecular Weight: 245.244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573504
CAS-Nummer: 220459-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR