2-Chloro-5-(trifluoromethoxy)pyridine 97%

Lieferant: Apollo Scientific
Achtung

PC200445-1G PC200445-5G
APOSPC200445-1GEA 144 EUR
APOSPC200445-1G APOSPC200445-5G
2-Chloro-5-(trifluoromethoxy)pyridine 97%
2-Chloro-5-(trifluoromethoxy)pyridine
Formel: C₆H₃ClF₃NO
Molecular Weight: 197,542 g/mol
Siedepunkt: 54 °C (15 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD18257007
CAS-Nummer: 1206972-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR