6-(Trifluoromethyl)-2-methylquinolin-4-ol
Lieferant: Apollo Scientific
PC200564-1G
PC200564-5G
APOSPC200564-1GEA
359
EUR
APOSPC200564-1G
APOSPC200564-5G
6-(Trifluoromethyl)-2-methylquinolin-4-ol
6-(Trifluoromethyl)-2-methylquinolin-4-ol
Formel:
C₁₁H₈F₃NO Molecular Weight: 227,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00272465 CAS-Nummer: 949507-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...