5-Chloro-3-fluoropyridin-2ylhydrazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-3-fluoro-2-hydrazinopyridine, 5-Chloro-3-fluoro-2-hydrazinylpyridine

PC2008-1G PC2008-5G
APOSPC2008-1GEA 81.2 EUR
APOSPC2008-1G APOSPC2008-5G
5-Chloro-3-fluoropyridin-2ylhydrazine
5-Chloro-3-fluoropyridin-2ylhydrazine
Formel: C₅H₅ClFN₃
Molecular Weight: 161.565 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09972183
CAS-Nummer: 248255-70-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR