5-Ethyl-2-(trifluoromethyl)pyridine 95+%

Lieferant: Apollo Scientific

PC201005-100MG PC201005-1G PC201005-250MG
APOSPC201005-100MGEA 81.2 EUR
APOSPC201005-100MG APOSPC201005-1G APOSPC201005-250MG
5-Ethyl-2-(trifluoromethyl)pyridine 95+%
5-Ethyl-2-(trifluoromethyl)pyridine
Formel: C₈H₈F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD19689817
CAS-Nummer: 1030632-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR