5-Cyclopropyl-2-fluoropyridine 97%
Lieferant: Apollo Scientific
Synonyme:
5-Cyclopropyl-2-flluoropyridine
PC201006-100MG
PC201006-1G
PC201006-250MG
APOSPC201006-100MGEA
168
EUR
APOSPC201006-100MG
APOSPC201006-1G
APOSPC201006-250MG
5-Cyclopropyl-2-fluoropyridine 97%
5-Cyclopropyl-2-fluoropyridine
Formel:
C₈H₈FN Molecular Weight: 137.154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18384289 CAS-Nummer: 1034467-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...