Methyl 6-fluoro-1H-indole-4-carboxylate 95%

Lieferant: Apollo Scientific

PC201020-1G PC201020-5G
APOSPC201020-1GEA 78.3 EUR
APOSPC201020-1G APOSPC201020-5G
Methyl 6-fluoro-1H-indole-4-carboxylate 95%
Methyl 6-fluoro-1H-indole-4-carboxylate
Formel: C₁₀H₈FNO₂
Molecular Weight: 193,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845444
CAS-Nummer: 1082040-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR