4-Chloro-2-fluoro-6-methoxybenzaldehyde 97%
Lieferant: Apollo Scientific
PC201036-100MG
PC201036-1G
PC201036-250MG
APOSPC201036-100MGEA
203
EUR
APOSPC201036-100MG
APOSPC201036-1G
APOSPC201036-250MG
4-Chloro-2-fluoro-6-methoxybenzaldehyde 97%
4-Chloro-2-fluoro-6-methoxybenzaldehyde
Formel:
C₈H₆ClFO₂ Molecular Weight: 188,583 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24686232 CAS-Nummer: 1158916-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...