2,2,2-Trifluoro-1-(p-tolyl)ethanamine hydrochloride 95+%

Lieferant: Apollo Scientific

PC201041-100MG PC201041-1G PC201041-250MG
APOSPC201041-100MGEA 121 EUR
APOSPC201041-100MG APOSPC201041-1G APOSPC201041-250MG
2,2,2-Trifluoro-1-(p-tolyl)ethanamine hydrochloride 95+%
2,2,2-Trifluoro-1-(p-tolyl)ethanamine hydrochloride
Formel: C₉H₁₀F₃N·ClH
Molecular Weight: 225.639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226565
CAS-Nummer: 1186195-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR