Methyl 4-bromo-2-fluoro-6-hydroxybenzoate 98%
Lieferant: Apollo Scientific
PC201047-100MG
PC201047-1G
PC201047-250MG
APOSPC201047-100MGEA
82.9
EUR
APOSPC201047-100MG
APOSPC201047-1G
APOSPC201047-250MG
Methyl 4-bromo-2-fluoro-6-hydroxybenzoate 98%
Methyl 4-bromo-2-fluoro-6-hydroxybenzoate
Formel:
C₈H₆BrFO₃ Molecular Weight: 249,034 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26391668 CAS-Nummer: 1193162-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...