trans-(4-(Trifluoromethyl)cyclohexyl)methanol 97%

Lieferant: Apollo Scientific
Achtung

PC201052-100MG PC201052-1G PC201052-250MG
APOSPC201052-100MGEA 98.9 EUR
APOSPC201052-100MG APOSPC201052-1G APOSPC201052-250MG
trans-(4-(Trifluoromethyl)cyclohexyl)methanol 97%
trans-(4-(Trifluoromethyl)cyclohexyl)methanol
Formel: C₈H₁₃F₃O
Molecular Weight: 182.183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29054589
CAS-Nummer: 1202577-61-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR