5-Bromo-6-fluoro-1H-pyrrolo[2,3-b]pyridine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC201055-100MG
PC201055-1G
PC201055-250MG
APOSPC201055-100MGEA
277
EUR
APOSPC201055-100MG
APOSPC201055-1G
APOSPC201055-250MG
5-Bromo-6-fluoro-1H-pyrrolo[2,3-b]pyridine 95%
5-Bromo-6-fluoro-1H-pyrrolo[2,3-b]pyridine
Formel:
C₇H₄BrFN₂ Molecular Weight: 215,022 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15529074 CAS-Nummer: 1207625-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...