5-Fluoro-2-methoxypyridin-3-amine 97%
Lieferant: Apollo Scientific
PC201061-100MG
PC201061-250MG
APOSPC201061-100MGEA
26.1
EUR
APOSPC201061-100MG
APOSPC201061-250MG
5-Fluoro-2-methoxypyridin-3-amine 97%
5-Fluoro-2-methoxypyridin-3-amine
Formel:
C₆H₇FN₂O Molecular Weight: 142.131 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18261779 CAS-Nummer: 1211541-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...