2-(Difluoromethoxy)pyridin-3-amine 97%

Lieferant: Apollo Scientific

PC201072-100MG PC201072-1G PC201072-250MG
APOSPC201072-100MGEA 119 EUR
APOSPC201072-100MG APOSPC201072-1G APOSPC201072-250MG
2-(Difluoromethoxy)pyridin-3-amine 97%
2-(Difluoromethoxy)pyridin-3-amine
Formel: C₆H₆F₂N₂O
Molecular Weight: 160,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185864
CAS-Nummer: 1214344-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR