3-(Chloromethyl)-1,1-difluorocyclobutane 95+%

Lieferant: Apollo Scientific

PC201113-100MG PC201113-1G PC201113-250MG
APOSPC201113-100MGEA 126 EUR
APOSPC201113-100MG APOSPC201113-1G APOSPC201113-250MG
3-(Chloromethyl)-1,1-difluorocyclobutane 95+%
3-(Chloromethyl)-1,1-difluorocyclobutane
Formel: C₅H₇ClF₂
Molecular Weight: 140.559 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19442317
CAS-Nummer: 1290625-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR