(S)-1-(3-Chloro-2-fluorophenyl)ethanamine hydrochloride 95+%

Lieferant: Apollo Scientific

PC201115-100MG PC201115-1G PC201115-250MG
APOSPC201115-100MGEA 179 EUR
APOSPC201115-100MG APOSPC201115-1G APOSPC201115-250MG
(S)-1-(3-Chloro-2-fluorophenyl)ethanamine hydrochloride 95+%
(S)-1-(3-Chloro-2-fluorophenyl)ethanamine hydrochloride
Formel: C₈H₉ClFN·ClH
Molecular Weight: 210.076 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12758016
CAS-Nummer: 1313593-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR