2,6-Dichloro-5-fluoro-4-methylnicotinic acid 95+%
Lieferant: Apollo Scientific
PC201119-100MG
PC201119-1G
PC201119-250MG
APOSPC201119-100MGEA
109
EUR
APOSPC201119-100MG
APOSPC201119-1G
APOSPC201119-250MG
2,6-Dichloro-5-fluoro-4-methylnicotinic acid 95+%
2,6-Dichloro-5-fluoro-4-methylnicotinic acid
Formel:
C₇H₄Cl₂FNO₂ Molecular Weight: 224,017 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044318 CAS-Nummer: 132195-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...