1-(3-Ethoxy-4-fluorophenyl)ethanone 97%

Lieferant: Apollo Scientific

PC201130-1G PC201130-250MG PC201130-5G
APOSPC201130-1GEA 275 EUR
APOSPC201130-1G APOSPC201130-250MG APOSPC201130-5G
1-(3-Ethoxy-4-fluorophenyl)ethanone 97%
1-(3-Ethoxy-4-fluorophenyl)ethanone
Formel: C₁₀H₁₁FO₂
Molecular Weight: 182,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375404
CAS-Nummer: 1378655-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR