2-Fluoro-1-(2-fluorophenyl)ethanone 95+%
Lieferant: Apollo Scientific
PC201134-100MG
PC201134-1G
PC201134-250MG
APOSPC201134-100MGEA
164
EUR
APOSPC201134-100MG
APOSPC201134-1G
APOSPC201134-250MG
2-Fluoro-1-(2-fluorophenyl)ethanone 95+%
2-Fluoro-1-(2-fluorophenyl)ethanone
Formel:
C₈H₆F₂O Molecular Weight: 156,129 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24713349 CAS-Nummer: 1402412-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...