8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 95+%

Lieferant: Apollo Scientific

PC201135-1G PC201135-5G
APOSPC201135-1GEA 72.5 EUR
APOSPC201135-1G APOSPC201135-5G
8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 95+%
8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one
Formel: C₁₁H₉FN₂O
Molecular Weight: 204.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381312
CAS-Nummer: 1408282-26-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR