4-Bromo-2-fluoro-6-hydroxybenzaldehyde 98%
Lieferant: Apollo Scientific
PC201140-100MG
PC201140-1G
PC201140-250MG
APOSPC201140-100MGEA
147
EUR
APOSPC201140-100MG
APOSPC201140-1G
APOSPC201140-250MG
4-Bromo-2-fluoro-6-hydroxybenzaldehyde 98%
4-Bromo-2-fluoro-6-hydroxybenzaldehyde
Formel:
C₇H₄BrFO₂ Molecular Weight: 219,008 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23705560 CAS-Nummer: 1427438-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...