1-Fluorocyclopentanecarboxylic acid 95+%
Lieferant: Apollo Scientific
PC201151-100MG
PC201151-250MG
PC201151-25MG
APOSPC201151-100MGEA
141
EUR
APOSPC201151-100MG
APOSPC201151-250MG
APOSPC201151-25MG
1-Fluorocyclopentanecarboxylic acid 95+%
1-Fluorocyclopentanecarboxylic acid
Formel:
C₆H₉FO₂ Molecular Weight: 132.133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26727072 CAS-Nummer: 1557370-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...