3-Fluoro-2-methylpyridin-4-amine 95+%

Lieferant: Apollo Scientific

PC201156-100MG PC201156-250MG PC201156-25MG
APOSPC201156-100MGEA 111 EUR
APOSPC201156-100MG APOSPC201156-250MG APOSPC201156-25MG
3-Fluoro-2-methylpyridin-4-amine 95+%
3-Fluoro-2-methylpyridin-4-amine
Formel: C₆H₇FN₂
Molecular Weight: 126.132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18261569
CAS-Nummer: 15931-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR