Ethyl 3-fluoro-1H-pyrrole-2-carboxylate 96%

Lieferant: Apollo Scientific
Achtung

PC201165-250MG PC201165-1G PC201165-5G
APOSPC201165-250MGEA 116 EUR
APOSPC201165-250MG APOSPC201165-1G APOSPC201165-5G
Ethyl 3-fluoro-1H-pyrrole-2-carboxylate 96%
Ethyl 3-fluoro-1H-pyrrole-2-carboxylate
Formel: C₇H₈FNO₂
Molecular Weight: 157,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042605
CAS-Nummer: 168102-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR