1-Chloro-2,5-difluoro-3-nitrobenzene 95+%

Lieferant: Apollo Scientific

PC201180-100MG PC201180-1G PC201180-250MG
APOSPC201180-100MGEA 49.3 EUR
APOSPC201180-100MG APOSPC201180-1G APOSPC201180-250MG
1-Chloro-2,5-difluoro-3-nitrobenzene 95+%
1-Chloro-2,5-difluoro-3-nitrobenzene
Formel: C₆H₂ClF₂NO₂
Molecular Weight: 193,535 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27923474
CAS-Nummer: 1805029-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR