2,4-Dichloro-5-fluoropyridine 98%

Lieferant: Apollo Scientific

PC201185-100MG PC201185-1G PC201185-250MG
APOSPC201185-100MGEA 119 EUR
APOSPC201185-100MG APOSPC201185-1G APOSPC201185-250MG
2,4-Dichloro-5-fluoropyridine 98%
2,4-Dichloro-5-fluoropyridine
Formel: C₅H₂Cl₂FN
Molecular Weight: 165,98 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18257011
CAS-Nummer: 189281-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR