1-Bromo-3,5-dimethoxy-4-fluorobenzene 95+%
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-2-fluoro-1,3-dimethoxybenzene
PC201193-1G
PC201193-25G
PC201193-5G
APOSPC201193-1GEA
111
EUR
APOSPC201193-1G
APOSPC201193-25G
APOSPC201193-5G
1-Bromo-3,5-dimethoxy-4-fluorobenzene 95+%
1-Bromo-3,5-dimethoxy-4-fluorobenzene
Formel:
C₈H₈BrFO₂ Molecular Weight: 235,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28738436 CAS-Nummer: 204654-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...