1-Chloro-6-fluorophthalazine 97%

Lieferant: Apollo Scientific

PC201211-100MG PC201211-1G PC201211-250MG
APOSPC201211-100MGEA 179 EUR
APOSPC201211-100MG APOSPC201211-1G APOSPC201211-250MG
1-Chloro-6-fluorophthalazine 97%
1-Chloro-6-fluorophthalazine
Formel: C₈H₄ClFN₂
Molecular Weight: 182.582 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432630
CAS-Nummer: 23928-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR