1-Chloro-7-fluorophthalazine 95+%

Lieferant: Apollo Scientific

PC201212-100MG PC201212-1G PC201212-250MG
APOSPC201212-100MGEA 215 EUR
APOSPC201212-100MG APOSPC201212-1G APOSPC201212-250MG
1-Chloro-7-fluorophthalazine 95+%
1-Chloro-7-fluorophthalazine
Formel: C₈H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18432628
CAS-Nummer: 23928-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR