Methyl 4-bromo-3-fluorothiophene-2-carboxylate 97%

Lieferant: Apollo Scientific

PC201246-100MG PC201246-250MG PC201246-1G
APOSPC201246-100MGEA 212 EUR
APOSPC201246-100MG APOSPC201246-250MG APOSPC201246-1G
Methyl 4-bromo-3-fluorothiophene-2-carboxylate 97%
Methyl 4-bromo-3-fluorothiophene-2-carboxylate
Formel: C₆H₄BrFO₂S
Molecular Weight: 239,062 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566744
CAS-Nummer: 395664-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR