6-Fluoro-1H-indazole-5-carbonitrile 95+%

Lieferant: Apollo Scientific

PC201272-100MG PC201272-250MG PC201272-1G
APOSPC201272-100MGEA 228 EUR
APOSPC201272-100MG APOSPC201272-250MG APOSPC201272-1G
6-Fluoro-1H-indazole-5-carbonitrile 95+%
6-Fluoro-1H-indazole-5-carbonitrile
Formel: C₈H₄FN₃
Molecular Weight: 161,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17010087
CAS-Nummer: 633327-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR