2-(3-Fluorophenyl)propan-2-amine hydrochloride 95+%

Lieferant: Apollo Scientific

PC201280-100MG PC201280-250MG PC201280-1G
APOSPC201280-100MGEA 78.3 EUR
APOSPC201280-100MG APOSPC201280-250MG APOSPC201280-1G
2-(3-Fluorophenyl)propan-2-amine hydrochloride 95+%
2-(3-Fluorophenyl)propan-2-amine hydrochloride
Formel: C₉H₁₂FN·ClH
Molecular Weight: 189.658 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870091
CAS-Nummer: 689232-61-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR