1-Bromo-2,5-difluoro-3-nitrobenzene 99%

Lieferant: Apollo Scientific
Achtung

PC201299-10G PC201299-1G PC201299-5G
APOSPC201299-10GEA 316 EUR
APOSPC201299-10G APOSPC201299-1G APOSPC201299-5G
1-Bromo-2,5-difluoro-3-nitrobenzene 99%
1-Bromo-2,5-difluoro-3-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18205960
CAS-Nummer: 741721-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR