(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine 95+%

Lieferant: Apollo Scientific

PC201306-100MG PC201306-250MG PC201306-1G
APOSPC201306-100MGEA 200 EUR
APOSPC201306-100MG APOSPC201306-250MG APOSPC201306-1G
(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine 95+%
(S)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine
Formel: C₈H₇BrF₃N
Molecular Weight: 254,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374593
CAS-Nummer: 778565-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR