2-Fluorobenzoyl cyanide 95+%
Lieferant: Apollo Scientific
PC201313-10G
PC201313-5G
PC201313-1G
PC201313-25G
APOSPC201313-10GEA
172
EUR
APOSPC201313-10G
APOSPC201313-5G
APOSPC201313-1G
APOSPC201313-25G
2-Fluorobenzoyl cyanide 95+%
2-Fluorobenzoyl cyanide
Formel:
C₈H₄FNO Molecular Weight: 149,122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658449 CAS-Nummer: 80277-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...