(R)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine 95+%

Lieferant: Apollo Scientific

PC201319-250MG PC201319-1G PC201319-5G
APOSPC201319-250MGEA 139 EUR
APOSPC201319-250MG APOSPC201319-1G APOSPC201319-5G
(R)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine 95+%
(R)-1-(4-Bromophenyl)-2,2,2-trifluoroethanamine
Formel: C₈H₇BrF₃N
Molecular Weight: 254,047 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374594
CAS-Nummer: 843608-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR