Methyl 2,6-difluoro-4-methoxybenzoate 95+%

Lieferant: Apollo Scientific

PC201322-10G PC201322-1G PC201322-5G
APOSPC201322-10GEA 748 EUR
APOSPC201322-10G APOSPC201322-1G APOSPC201322-5G
Methyl 2,6-difluoro-4-methoxybenzoate 95+%
Methyl 2,6-difluoro-4-methoxybenzoate
Formel: C₉H₈F₂O₃
Molecular Weight: 202,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10566175
CAS-Nummer: 84937-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR