7-Fluoro-1H-indole-3-carboxylic acid 98%
Lieferant: Apollo Scientific
PC201326-250MG
PC201326-1G
APOSPC201326-250MGEA
239
EUR
APOSPC201326-250MG
APOSPC201326-1G
7-Fluoro-1H-indole-3-carboxylic acid 98%
7-Fluoro-1H-indole-3-carboxylic acid
Formel:
C₉H₆FNO₂ Molecular Weight: 179,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09954940 CAS-Nummer: 858515-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...