(S)-1-(3-Fluoro-4-methoxyphenyl)ethanamine 95+%

Lieferant: Apollo Scientific

PC201337-100MG PC201337-1G PC201337-250MG
APOSPC201337-100MGEA 224 EUR
APOSPC201337-100MG APOSPC201337-1G APOSPC201337-250MG
(S)-1-(3-Fluoro-4-methoxyphenyl)ethanamine 95+%
(S)-1-(3-Fluoro-4-methoxyphenyl)ethanamine
Formel: C₉H₁₂FNO·ClH
Molecular Weight: 205,657 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06761862
CAS-Nummer: 870849-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR