5-Fluoro-6-methoxynicotinaldehyde 98%

Lieferant: Apollo Scientific

PC201349-250MG PC201349-1G
APOSPC201349-250MGEA 187 EUR
APOSPC201349-250MG APOSPC201349-1G
5-Fluoro-6-methoxynicotinaldehyde 98%
5-Fluoro-6-methoxynicotinaldehyde
Formel: C₇H₆FNO₂
Molecular Weight: 155,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112587
CAS-Nummer: 884494-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR