6-Fluoro-1H-indol-4-amine 97%

Lieferant: Apollo Scientific

PC201356-100MG PC201356-250MG PC201356-1G
APOSPC201356-100MGEA 144 EUR
APOSPC201356-100MG APOSPC201356-250MG APOSPC201356-1G
6-Fluoro-1H-indol-4-amine 97%
6-Fluoro-1H-indol-4-amine
Formel: C₈H₇FN₂
Molecular Weight: 150.153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07357294
CAS-Nummer: 885518-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR