5-Fluoro-2-methylisonicotinic acid 97%

Lieferant: Apollo Scientific

PC201357-100MG PC201357-250MG PC201357-1G
APOSPC201357-100MGEA 257 EUR
APOSPC201357-100MG APOSPC201357-250MG APOSPC201357-1G
5-Fluoro-2-methylisonicotinic acid 97%
5-Fluoro-2-methylisonicotinic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18256823
CAS-Nummer: 885588-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR