(4-Bromo-2,6-difluorophenyl)methanamine 97%

Lieferant: Apollo Scientific

PC201363-250MG PC201363-1G PC201363-5G
APOSPC201363-250MGEA 40.6 EUR
APOSPC201363-250MG APOSPC201363-1G APOSPC201363-5G
(4-Bromo-2,6-difluorophenyl)methanamine 97%
(4-Bromo-2,6-difluorophenyl)methanamine
Formel: C₇H₆BrF₂N
Molecular Weight: 222.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972477
CAS-Nummer: 887585-99-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR