6-Chloro-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 95+%

Lieferant: Apollo Scientific

PC201378-100MG PC201378-250MG
APOSPC201378-100MGEA 203 EUR
APOSPC201378-100MG APOSPC201378-250MG
6-Chloro-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine 95+%
6-Chloro-3-(trifluoromethyl)-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₄ClF₃N₂
Molecular Weight: 220,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189868
CAS-Nummer: 932406-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR