5,6-Difluoroindoline 97%

Lieferant: Apollo Scientific

PC201385-100MG PC201385-250MG PC201385-1G
APOSPC201385-100MGEA 58 EUR
APOSPC201385-100MG APOSPC201385-250MG APOSPC201385-1G
5,6-Difluoroindoline 97%
5,6-Difluoroindoline
Formel: C₈H₇F₂N
Molecular Weight: 155,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01075221
CAS-Nummer: 954255-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR